APO455838920 Apollo Scientific Ltd

2-Fluoro-3-methylbenzaldehyde

  • MDL number MFCD06797948
  • Cas Number : 886762-64-7
  • Molecular Weight : 138.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-m-tolualdehyde, 2-Fluoro-3-formyltoluene
Flash Point 79
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 77-79/10mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-methylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-methylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart