APO455838924 Apollo Scientific Ltd

4-Iodo-3-(trifluoromethoxy)benzoic acid

  • MDL number MFCD07368785
  • Cas Number : 886762-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9206

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=9206
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart