APO455838926 Apollo Scientific Ltd

3-Fluoro-4-formylbenzonitrile

  • MDL number MFCD07368786
  • Cas Number : 105942-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-79
Purity 98
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-formylbenzonitrile-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-formylbenzonitrile-98
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart