APO455838928 Apollo Scientific Ltd

2-Bromo-6-fluoronitrobenzene

  • MDL number MFCD07368788
  • Cas Number : 886762-70-5
  • Molecular Weight : 220

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-37
Synonym 1-Bromo-3-fluoro-2-nitrobenzene
Purity 99
Flash Point 87
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoronitrobenzene
pictograms GHS07
hstatements H302;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart