APO455838929 Apollo Scientific Ltd

2,3-Dibromofluorobenzene

  • MDL number MFCD07368789
  • Cas Number : 811711-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 15-20
Synonym 1,2-Dibromo-3-fluorobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/23-dibromofluorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-dibromofluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart