APO455838930 Apollo Scientific Ltd

2-Fluoro-6-iodonitrobenzene

  • MDL number MFCD07368790
  • Cas Number : 886762-71-6
  • Molecular Weight : 267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-37
Synonym 1-Fluoro-3-iodo-2-nitrobenzene
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-iodonitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-iodonitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart