APO455838948 Apollo Scientific Ltd

3-Fluoro-4-nitrobenzonitrile

  • MDL number MFCD04973417
  • Cas Number : 218632-01-0
  • Molecular Weight : 166.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-75
Synonym 4-Cyano-2-fluoronitrobenzene
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-nitrobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-nitrobenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart