APO455838957 Apollo Scientific Ltd

5-(2,4-Difluorophenyl)isoxazole

  • MDL number MFCD02183538
  • Cas Number : 138716-46-8
  • Molecular Weight : 181.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-24-difluorophenylisoxazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-24-difluorophenylisoxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart