APO455838963 Apollo Scientific Ltd

3',5'-Difluoroacetanilide

  • MDL number MFCD03094568
  • Cas Number : 404-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3009

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3009
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart