APO455838967 Apollo Scientific Ltd

4-Chloro-3-fluorotoluene

  • MDL number MFCD00143295
  • Cas Number : 5527-94-6
  • Molecular Weight : 144.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.2
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 157-158
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-fluorotoluene

Safety Information

signalword Warning
pcodes P210;P280;P242
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-fluorotoluene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart