APO455838973 Apollo Scientific Ltd

N-(Chloroacetyl)-2-fluoroaniline

  • MDL number MFCD00730970
  • Cas Number : 347-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-2'-fluoroacetanilide
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/n-chloroacetyl-2-fluoroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/n-chloroacetyl-2-fluoroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart