APO456788952 Apollo Scientific Ltd

3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one

  • MDL number MFCD06245367
  • Cas Number : 178445-83-5
  • Molecular Weight : 286.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-34-dimethoxyphenyl-1-3-hydroxyphenylpropan-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-34-dimethoxyphenyl-1-3-hydroxyphenylpropan-1-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart