APO456789314 Apollo Scientific Ltd

(Morpholin-4-yl)(piperazin-1-yl)methanone

  • MDL number MFCD03844725
  • Cas Number : 98834-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-88
Synonym 1-[(Morpholin-4-yl)carbonyl]piperazine, 4-[(Piperazin-1-yl)carbonyl]morpholine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28411

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28411
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart