APO456789357 Apollo Scientific Ltd

5-Amino-3-methylcinnoline

  • MDL number MFCD00466524
  • Cas Number : 300690-74-8
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 189-191
Iupac Name 3-methyl-5-cinnolinylamine
Inchi Code 1S/C9H9N3/c1-6-5-7-8(10)3-2-4-9(7)12-11-6/h2-5H,10H2,1H3
Inchi Key HOJQDQQMHMVETF-UHFFFAOYSA-N

Description

Synonym 3-Methylcinnolin-5-amine, 5-Amino-3-methyl-1,2-benzodiazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 189-191
Product Link https://store.apolloscientific.co.uk/product/5-amino-3-methylcinnoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-3-methylcinnoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart