APO456789405 Apollo Scientific Ltd

Ethyl oct-7-enoate

  • MDL number MFCD00671846
  • Cas Number : 35194-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-oct-7-enoate

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-oct-7-enoate
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart