APO456789434 Apollo Scientific Ltd

1-Allyl-4-chlorobenzene

  • MDL number MFCD01319547
  • Cas Number : 1745-18-2
  • Molecular Weight : 152.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-4-(prop-2-en-1-yl)benzene, 3-(4-Chlorophenyl)prop-1-ene
Density 1.05
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 199-201
Product Link https://store.apolloscientific.co.uk/product/1-allyl-4-chlorobenzene

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-allyl-4-chlorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart