APO456789491 Apollo Scientific Ltd

5-Chloro-2-methoxyphenyl isocyanate

  • MDL number MFCD00013860
  • Cas Number : 55440-54-5
  • Molecular Weight : 183.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-62
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-methoxyphenyl-isocyanate

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-methoxyphenyl-isocyanate
pictograms GHS07;GHS08
hstatements EUH208;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart