APO456789564 Apollo Scientific Ltd

Methyl 1,2,3-benzothiadiazole-5-carboxylate

  • MDL number MFCD01566550
  • Cas Number : 23616-15-1
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Synonym 5-(Methoxycarbonyl)-1,2,3-benzothiadiazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-123-benzothiadiazole-5-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-123-benzothiadiazole-5-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart