APO456789603 Apollo Scientific Ltd

3-(Piperidin-1-yl)benzylamine

  • MDL number MFCD06659082
  • Cas Number : 175696-71-6
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-piperidinyl)phenyl]methanamine
Inchi Code 1S/C12H18N2/c13-10-11-5-4-6-12(9-11)14-7-2-1-3-8-14/h4-6,9H,1-3,7-8,10,13H2
Inchi Key BISSHLVDQOLHNQ-UHFFFAOYSA-N

Description

Synonym [3-(Piperidin-1-yl)phenyl]methylamine, 1-[3-(Aminomethyl)phenyl]piperidine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid-Viscous
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-piperidin-1-ylbenzylamine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-piperidin-1-ylbenzylamine
pictograms GHS05
hstatements H314;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart