APO456789737 Apollo Scientific Ltd

1,2-Thiazinane 1,1-dioxide

  • MDL number MFCD04038916
  • Cas Number : 37441-50-2
  • Molecular Weight : 135.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-96
Synonym 1,1-Dioxotetrahydro-2H-1,2-thiazine, 1,4-Butanesultam
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/12-thiazinane-11-dioxide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/12-thiazinane-11-dioxide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart