APO456789774 Apollo Scientific Ltd

7-(Hydroxymethyl)-1H-indole

  • MDL number MFCD02179597
  • Cas Number : 1074-87-9
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-48
Synonym (1H-Indol-7-yl)methanol
Purity 97
Storage temp. Store at 2-8
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-hydroxymethyl-1h-indole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/7-hydroxymethyl-1h-indole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart