APO456789875 Apollo Scientific Ltd

5-Methoxy-1-methyl-1H-indole-3-carboxaldehyde

  • MDL number MFCD01578324
  • Cas Number : 39974-94-2
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 126-129
Iupac Name 5-methoxy-1-methyl-1H-indole-3-carbaldehyde
Inchi Code 1S/C11H11NO2/c1-12-6-8(7-13)10-5-9(14-2)3-4-11(10)12/h3-7H,1-2H3
Inchi Key OFNAEHQJIMREFJ-UHFFFAOYSA-N

Description

Synonym 3-Formyl-5-methoxy-1-methyl-1H-indole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 126-129
Product Link https://store.apolloscientific.co.uk/product/5-methoxy-1-methyl-1h-indole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methoxy-1-methyl-1h-indole-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart