APO456789889 Apollo Scientific Ltd

1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-amine

  • MDL number MFCD11505007
  • Cas Number : 1190380-49-4
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Synonym 4-Amino-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazole, 4-(4-Amino-1H-pyrazol-1-yl)tetrahydro-2H-pyran
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart