APO456789891 Apollo Scientific Ltd

4,5,6,7-Tetrahydrobenzo[b]thiophene

  • MDL number MFCD19442759
  • Cas Number : 13129-17-4
  • Molecular Weight : 138.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5,6,7-Tetrahydro-1-benzothiophene
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 52-53/3mm
Product Link https://store.apolloscientific.co.uk/product/4567-tetrahydrobenzobthiophene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4567-tetrahydrobenzobthiophene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart