APO456789922 Apollo Scientific Ltd

5-Amino-4-phenyl-1,2,3-thiadiazole

  • MDL number MFCD00100232
  • Cas Number : 78301-72-1
  • Molecular Weight : 177.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 146-147
Iupac Name 4-phenyl-1,2,3-thiadiazol-5-amine
Inchi Code 1S/C8H7N3S/c9-8-7(10-11-12-8)6-4-2-1-3-5-6/h1-5H,9H2
Inchi Key VPHYLVGCMWSAEJ-UHFFFAOYSA-N

Description

Synonym 4-Phenyl-1,2,3-thiadiazol-5-amine, (5-Amino-1,2,3-thiadiazol-4-yl)benzene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 146-147
Product Link https://store.apolloscientific.co.uk/product/5-amino-4-phenyl-123-thiadiazole

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-4-phenyl-123-thiadiazole
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart