APO456789924 Apollo Scientific Ltd

2-(Bromoacetyl)furan

  • MDL number MFCD04038962
  • Cas Number : 15109-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32-35
Synonym 2-Bromo-1-(fur-2-yl)ethan-1-one, 2-Bromo-1-(fur-2-yl)-1-oxoethane
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Boiling point 72-74/0.2mm
Product Link https://store.apolloscientific.co.uk/product/2-bromoacetylfuran

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromoacetylfuran
pictograms GHS05;GHS07
hstatements H314;H302+H332
Availability details for
Quantity
  • Availability
  • Add to Cart