APO456789981 Apollo Scientific Ltd

1-Bromo-2-(methoxymethyl)benzene

  • MDL number MFCD03790887
  • Cas Number : 52711-30-5
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromobenzyl methyl ether
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 75-80/0.5mm
Product Link https://store.apolloscientific.co.uk/product/1-bromo-2-methoxymethylbenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-2-methoxymethylbenzene
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart