APO456790025 Apollo Scientific Ltd

4-Chloro-2-methoxyaniline

  • MDL number MFCD06801386
  • Cas Number : 93-50-5
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-53
Synonym 2-Amino-5-chloroanisole
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 260
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-methoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-methoxyaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart