APO456790289 Apollo Scientific Ltd

N-(2-Chloroethyl)-N'-(3-chloro-4-methylphenyl)urea

  • MDL number MFCD08689727
  • Cas Number : 937601-77-9
  • Molecular Weight : 247.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-148
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-2-chloroethyl-n-3-chloro-4-methylphenylurea

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/n-2-chloroethyl-n-3-chloro-4-methylphenylurea
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart