APO456790419 Apollo Scientific Ltd

{4-[(Tetrahydro-2H-pyran-4-yl)oxy]phenyl}methylamine

  • MDL number MFCD09064967
  • Cas Number : 864266-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41.5-43.5
Synonym 4-[(Tetrahydro-2H-pyran-4-yl)oxy]benzylamine, 4-[4-(Aminomethyl)phenoxy]tetrahydro-2H-pyran
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37831

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37831
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart