APO456790490 Apollo Scientific Ltd

Benzo[b]thiophene-7-carboxylic acid

  • MDL number MFCD08572208
  • Cas Number : 10134-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176.5-178.5
Synonym 1-Benzothiophene-7-carboxylic acid, 7-Carboxybenzo[b]thiophene
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/benzobthiophene-7-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/benzobthiophene-7-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart