APO456790556 Apollo Scientific Ltd

1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxaldehyde

  • MDL number MFCD09817540
  • Cas Number : 916766-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Synonym 4-Formyl-1-(thieno[3,2-d]pyrimidin-4-yl)piperidine, 4-(4-Formylpiperidin-1-yl)thieno[3,2-d]pyrimidine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38136

Safety Information

signalword Warning
pcodes P261;P271;P281
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38136
pictograms GHS07
hstatements H302;H312;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart