APO456790608 Apollo Scientific Ltd

2-(3,5-Dichlorophenyl)pyrrolidine

  • MDL number MFCD02684093
  • Cas Number : 887344-13-0
  • Molecular Weight : 216.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,5-dichlorophenyl)pyrrolidine
Inchi Code 1S/C10H11Cl2N/c11-8-4-7(5-9(12)6-8)10-2-1-3-13-10/h4-6,10,13H,1-3H2
Inchi Key RGJUGOHYPAYSDI-UHFFFAOYSA-N

Description

Synonym 2-(3,5-Dichlorophenyl)tetrahydropyrrole, 1,3-Dichloro-5-(pyrrolidin-2-yl)benzene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-35-dichlorophenylpyrrolidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-35-dichlorophenylpyrrolidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart