APO456790615 Apollo Scientific Ltd

4-(Pyrrolidin-2-yl)pyridine

  • MDL number MFCD01862536
  • Cas Number : 128562-25-4
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Pyridin-4-yl)pyrrolidine
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-pyrrolidin-2-ylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-pyrrolidin-2-ylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart