APO456790645 Apollo Scientific Ltd

4-(2-Chlorophenyl)benzylamine hydrochloride

  • MDL number MFCD02089431
  • Cas Number : 518357-42-1
  • Molecular Weight : 254.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2'-chloro[1,1'-biphenyl]-4-yl)methanamine hydrochloride
Inchi Code 1S/C13H12ClN.ClH/c14-13-4-2-1-3-12(13)11-7-5-10(9-15)6-8-11;/h1-8H,9,15H2;1H
Inchi Key URGRCECNVGEABW-UHFFFAOYSA-N

Description

Synonym [4-(2-Chlorophenyl)phenyl]methylamine hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-2-chlorophenylbenzylamine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/4-2-chlorophenylbenzylamine-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart