APO456790764 Apollo Scientific Ltd

trans-3-Phenoxycinnamic acid

  • MDL number MFCD00736661
  • Cas Number : 77124-20-0
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 114-115
Iupac Name (2E)-3-(3-phenoxyphenyl)-2-propenoic acid
Inchi Code 1S/C15H12O3/c16-15(17)10-9-12-5-4-8-14(11-12)18-13-6-2-1-3-7-13/h1-11H,(H,16,17)/b10-9+
Inchi Key RYTBBLWPDBZDHT-MDZDMXLPSA-N

Description

Synonym (E)-3-(3-Phenoxyphenyl)acrylic acid, (2E)-3-(3-Phenoxyphenyl)prop-2-enoic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 114-115
Product Link https://store.apolloscientific.co.uk/product/trans-3-phenoxycinnamic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-3-phenoxycinnamic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart