APO456790771 Apollo Scientific Ltd

5-Amino-2-phenyl-2H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD02218166
  • Cas Number : 103752-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Amino-5-carbamoyl-2-phenyl-2H-1,2,3-triazole, (4-Amino-5-carbamoyl-2H-1,2,3-triazol-2-yl)benzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38610

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38610
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart