APO456790802 Apollo Scientific Ltd

8-Amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD09998375
  • Cas Number : 870064-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94
Synonym 8-Amino-3,4-dihydro-2,4-dimethyl-3-oxo-2H-1,4-benzoxazine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42971

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42971
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart