APO456790918 Apollo Scientific Ltd

Methyl 2-(chloromethyl)-1,3-benzoxazole-5-carboxylate

  • MDL number MFCD08689722
  • Cas Number : 924862-18-0
  • Molecular Weight : 225.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 90-91
Iupac Name methyl 2-(chloromethyl)-1,3-benzoxazole-5-carboxylate
Inchi Code 1S/C10H8ClNO3/c1-14-10(13)6-2-3-8-7(4-6)12-9(5-11)15-8/h2-4H,5H2,1H3
Inchi Key RQZPYCDYMVRMSC-UHFFFAOYSA-N

Description

Synonym 2-(Chloromethyl)-5-(methoxycarbonyl)-1,3-benzoxazole, 2-(Chloromethyl)-5-(methoxycarbonyl)benzo[d]oxazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 90-91
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloromethyl-13-benzoxazole-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloromethyl-13-benzoxazole-5-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart