APO456790927 Apollo Scientific Ltd

4-(Isopropoxycarbonyl)benzeneboronic acid

  • MDL number MFCD02259385
  • Cas Number : 342002-82-8
  • Molecular Weight : 208.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111(dec.)
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-isopropoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-isopropoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart