APO456791020 Apollo Scientific Ltd

2-(1H-Pyrrol-1-yl)benzonitrile

  • MDL number MFCD00128308
  • Cas Number : 33265-71-3
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Synonym 1-(2-Cyanophenyl)-1H-pyrrole
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-1h-pyrrol-1-ylbenzonitrile

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-1h-pyrrol-1-ylbenzonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart