APO456791032 Apollo Scientific Ltd

2-(Hydroxymethyl)chroman

  • MDL number MFCD06658990
  • Cas Number : 83278-86-8
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-dihydro-2H-chromen-2-ylmethanol
Inchi Code 1S/C10H12O2/c11-7-9-6-5-8-3-1-2-4-10(8)12-9/h1-4,9,11H,5-7H2
Inchi Key KDLVSGWUKFJFTL-UHFFFAOYSA-N

Description

Synonym (3,4-Dihydro-2H-chromen-2-yl)methanol, (3,4-Dihydro-2H-1-benzopyran-2-yl)methanol
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxymethylchroman

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxymethylchroman
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart