APO456791123 Apollo Scientific Ltd

7-(Aminomethyl)-1-methyl-1H-indole

  • MDL number MFCD09879961
  • Cas Number : 937795-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1-Methyl-1H-indol-7-yl)methylamine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42201

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42201
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart