APO456791164 Apollo Scientific Ltd

Methyl 3-amino-6-bromobenzo[b]thiophene-2-carboxylate

  • MDL number MFCD09865017
  • Cas Number : 1017782-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-128
Synonym Methyl 3-amino-6-bromo-1-benzothiophene-2-carboxylate,3-Amino-6-bromo-2-(methoxycarbonyl)benzo[b]thiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-3-amino-6-bromobenzobthiophene-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-3-amino-6-bromobenzobthiophene-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart