APO456791168 Apollo Scientific Ltd

5-Methyl-3-nitro-1H-pyrazole

  • MDL number MFCD00514181
  • Cas Number : 34334-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-methyl-3-nitro-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-3-nitro-1h-pyrazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart