APO456791300 Apollo Scientific Ltd

Potassium 4-iodobenzenesulphonate

  • MDL number MFCD06656232
  • Cas Number : 13035-63-7
  • Molecular Weight : 322.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name potassium 4-iodobenzenesulfonate
Inchi Code 1S/C6H5IO3S.K/c7-5-1-3-6(4-2-5)11(8,9)10;/h1-4H,(H,8,9,10);/q;+1/p-1
Inchi Key JKXILXFUIWOQAP-UHFFFAOYSA-M

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/potassium-4-iodobenzenesulphonate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/potassium-4-iodobenzenesulphonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart