APO456791304 Apollo Scientific Ltd

Thieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD07371381
  • Cas Number : 59944-76-2
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 293-294
Synonym 2-Carboxythieno[2,3-b]pyridine, 2-Carboxy-7-azabenzo[b]thiophene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/thieno23-bpyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/thieno23-bpyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart