APO456791325 Apollo Scientific Ltd

4-Amino-3-methylbenzenesulphonamide

  • MDL number MFCD00466840
  • Cas Number : 53297-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160
Synonym 4-(Aminosulphonyl)-2-methylaniline, 2-Amino-5-(aminosulphonyl)toluene
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-methylbenzenesulphonamide

Safety Information

signalword Warning
pcodes P280;P271
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-methylbenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart