APO456791402 Apollo Scientific Ltd

4-[(tert-Butyl)sulphanyl]benzaldehyde

  • MDL number MFCD08443944
  • Cas Number : 88357-16-8
  • Molecular Weight : 194.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[(tert-Butyl)thio]benzaldehyde, tert-Butyl 4-formylphenyl sulphide
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butylsulphanylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butylsulphanylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart