APO456791476 Apollo Scientific Ltd

Methyl 4-chlorobenzo[b]thiophene-2-carboxylate

  • MDL number MFCD07371546
  • Cas Number : 35212-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-4-chlorobenzobthiophene-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-chlorobenzobthiophene-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart